Learn More
Enterprise Technology Solutions Clorox Company Hand Sanitizer, Bleach-Free, 1L, 6/CT, Clear
SDP
Catalog No. 5000790472
Encompass_Preferred
The Clorox Hand Sanitizer 1 Liter Refill unit for a push-button dispenser is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. Hand sanitizer is great for use in healthcare, foodservice and educational institutions. From Clorox Commercial Solutions, this hand sanitizer refill works with the wall-mounted, push-button, Clorox Hand Sanitizer Dispenser unit (not included).
Catalog No. 50-007-90472 Supplier Enterprise Technology Solutions Supplier No. CLO01753
The Clorox Hand Sanitizer 1 Liter Refill unit for a push-button dispenser is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. Hand sanitizer is great for use in healthcare, foodservice and educational institutions. From Clorox Commercial Solutions, this hand sanitizer refill works with the wall-mounted, push-button, Clorox Hand Sanitizer Dispenser unit (not included).