Learn More
Enterprise Technology Solutions Clorox Company Hand Sanitizer, Bleach-free, 2 oz. Spray Bottle, 5208/PL, CL
SDP
Catalog No. 5000790438
Encompass_Preferred
Clorox Hand Sanitizer is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. From Clorox Commercial Solutions, the small, 2 oz. size is perfect for on-the-go use.
Catalog No. 50-007-90438 Supplier Enterprise Technology Solutions Supplier No. CLO02174PL
Clorox Hand Sanitizer is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. From Clorox Commercial Solutions, the small, 2 oz. size is perfect for on-the-go use.