Learn More
Enterprise Technology Solutions Clorox Company Scrub Pads w/Soap,Heavy-Duty,Steel Wool,5"x4",15/BX,240BX/BD
SDP
Catalog No. 5000831654
Encompass_Preferred
S.O.S Steel Wool Soap Pads are tough so you don't have to be. Made with durable steel wool and saturated with long-lasting soap, these pads cut through even the toughest, baked-on, greasy, grimy messes. They are perfect for cleaning ovens, broilers, cookie sheets and pots and pans. These reusable, soap pads are great outside the kitchen too. From scrubbing stoves and shower doors to cleaning grills and golf clubs, they quickly remove stubborn grease, rust and stains inside and outside the home or workplace. The soap contains rust inhibitors, preservatives, soaps and detergents, a pH buffer, fragrance and color. From CloroxPro, these steel wool soap pads are ideal for use in restaurants, cafeterias, hotels, breakrooms and other commercial facilities. Cleaning is quick and easy with S.O.S.
Catalog No. 50-008-31654 Supplier Enterprise Technology Solutions Supplier No. CLO88320BD
S.O.S Steel Wool Soap Pads are tough so you don't have to be. Made with durable steel wool and saturated with long-lasting soap, these pads cut through even the toughest, baked-on, greasy, grimy messes. They are perfect for cleaning ovens, broilers, cookie sheets and pots and pans. These reusable, soap pads are great outside the kitchen too. From scrubbing stoves and shower doors to cleaning grills and golf clubs, they quickly remove stubborn grease, rust and stains inside and outside the home or workplace. The soap contains rust inhibitors, preservatives, soaps and detergents, a pH buffer, fragrance and color. From CloroxPro, these steel wool soap pads are ideal for use in restaurants, cafeterias, hotels, breakrooms and other commercial facilities. Cleaning is quick and easy with S.O.S.