Learn More
Enterprise Technology Solutions Clorox Company Scrubber Sponges, All Surface, 2-1/2"x4-1/2", 2100/BD, BE
SDP
Catalog No. 5000830994
Encompass_Preferred
S.O.S All Surface Scrubber Sponge is dual-sided for cleaning both heavily soiled and delicate surfaces. It's durable and long-lasting for everyday use with the scrubbing power on one side to cut through tough messes and a soft, absorbent cellulose sponge on the other to clean delicate surfaces including nonstick cookware, marble, ceramic and tile. The all-surface sponge features an easy-to-hold shape that allows you to clean hard-to-reach crevices faster and easier. This sponge is also dishwasher-safe so you can toss it in the top rack to conveniently sanitize. It is ideal for use in restaurants, cafeterias, hotels, breakrooms and other commercial facilities. Cleaning is quick and easy with S.O.S.
Catalog No. 50-008-30994 Supplier Enterprise Technology Solutions Supplier No. CLO91017BD
S.O.S All Surface Scrubber Sponge is dual-sided for cleaning both heavily soiled and delicate surfaces. It's durable and long-lasting for everyday use with the scrubbing power on one side to cut through tough messes and a soft, absorbent cellulose sponge on the other to clean delicate surfaces including nonstick cookware, marble, ceramic and tile. The all-surface sponge features an easy-to-hold shape that allows you to clean hard-to-reach crevices faster and easier. This sponge is also dishwasher-safe so you can toss it in the top rack to conveniently sanitize. It is ideal for use in restaurants, cafeterias, hotels, breakrooms and other commercial facilities. Cleaning is quick and easy with S.O.S.