Learn More
Enterprise Technology Solutions Clorox Company Wipes, Free&Clear, 75 Wipes, 4-1/4"x4-1/4"x8-3/10", White
SDP
Catalog No. 5000790830
Encompass_Preferred
Safe around children, pets and food-contact surfaces when used as directed, Clorox Free and Clear Compostable Wipes are formulated with no dyes, bleach or ammonia yet deliver a powerful clean that's gentle on surfaces. Fragrance-free wipes pick up dirt and can compost back into dirt. They are tough on grease and cut through grime on a variety of hard surfaces such as acrylic, sealed fiberglass, sealed granite, finished hardwood and vinyl. They meet U.S. EPA's Safer Choice product standards and are made from plant-based fibers that are compostable under home conditions only when used as directed. Do not compost wipes if used to clean noncompostable messes such as human or animal waste. Wipes should be no more than 10 percent of your total home compost pile. Grab these easy, accessible cleaning wipes for everyday cleanups and tough messes. Do not flush.
Catalog No. 50-007-90830 Supplier Enterprise Technology Solutions Supplier No. CLO32486
Safe around children, pets and food-contact surfaces when used as directed, Clorox Free and Clear Compostable Wipes are formulated with no dyes, bleach or ammonia yet deliver a powerful clean that's gentle on surfaces. Fragrance-free wipes pick up dirt and can compost back into dirt. They are tough on grease and cut through grime on a variety of hard surfaces such as acrylic, sealed fiberglass, sealed granite, finished hardwood and vinyl. They meet U.S. EPA's Safer Choice product standards and are made from plant-based fibers that are compostable under home conditions only when used as directed. Do not compost wipes if used to clean noncompostable messes such as human or animal waste. Wipes should be no more than 10 percent of your total home compost pile. Grab these easy, accessible cleaning wipes for everyday cleanups and tough messes. Do not flush.