Learn More
NDC Inc 121oz. Clorox Germicidal Bleach
Catalog No. 50371500
Encompass_Preferred
New 121oz. Clorox Germicidal Bleach, with a concentration level of 8.25%, is EPA-registered to kill more organisms than original Ultra Clorox Germicidal Bleach as well as private label germicidal bleach. With the dosage requirements for general disinfection cut in half, Clorox Germicidal Bleach provides better value when comparing number of uses per case for general disinfection.
Catalog No. 50371500 Supplier NDC Inc Supplier No. CLO30966
New 121oz. Clorox Germicidal Bleach, with a concentration level of 8.25%, is EPA-registered to kill more organisms than original Ultra Clorox Germicidal Bleach as well as private label germicidal bleach. With the dosage requirements for general disinfection cut in half, Clorox Germicidal Bleach provides better value when comparing number of uses per case for general disinfection.