Learn More
Andwin Scientific CLOROX WPS 150/CANISTER 8/CS
SDP
Catalog No. NC0247830
Encompass_Preferred
The Dispatch hospital cleaner towels are efficient disinfectant for countertop surfaces, carts, and equipments. This product is ready-to-use as it is already premixed and stabilized. In 1 minute, this product can kill a wide range of viruses and fungus and within 30 seconds, it can eliminate TB (mycobacterium bovis) bacterium. This product is packed in a canister and is sold in a case of 8.
This item is not returnable. View return policy
Catalog No. NC0247830 Supplier Andwin Scientific Supplier No. 89042154
The Dispatch hospital cleaner towels are efficient disinfectant for countertop surfaces, carts, and equipments. This product is ready-to-use as it is already premixed and stabilized. In 1 minute, this product can kill a wide range of viruses and fungus and within 30 seconds, it can eliminate TB (mycobacterium bovis) bacterium. This product is packed in a canister and is sold in a case of 8.