Learn More
Grainger CLOROX HEALTHCARE Germicidal Disinfecting Wipes
Catalog No. NC2204166
Encompass_Preferred
Clorox Healthcare Germicidal Disinfecting Wipes are EPA-registered disinfecting wipes that kill 99.9% of bacteria and viruses including SARS-CoV-2, the virus that causes COVID-19. These pre-moistened wipes come in canisters of 160 wipes that are 6" x 10" in size. They are bleach-free, fragrance-free, and made of strong cloth that will not tear easily. The wipes are safe for use on hard, nonporous surfaces and are ideal for disinfecting high-touch surfaces in healthcare settings or other facilities. Each wipe has a 4 minute contact time for disinfecting against pathogens. These wipes meet CDC guidelines for disinfecting.
This item is not returnable. View return policy
Catalog No. NC2204166 Supplier Grainger Supplier No. 6VDE9
Clorox Healthcare Germicidal Disinfecting Wipes are EPA-registered disinfecting wipes that kill 99.9% of bacteria and viruses including SARS-CoV-2, the virus that causes COVID-19. These pre-moistened wipes come in canisters of 160 wipes that are 6" x 10" in size. They are bleach-free, fragrance-free, and made of strong cloth that will not tear easily. The wipes are safe for use on hard, nonporous surfaces and are ideal for disinfecting high-touch surfaces in healthcare settings or other facilities. Each wipe has a 4 minute contact time for disinfecting against pathogens. These wipes meet CDC guidelines for disinfecting.