Learn More
Glas Col LLC Glove Bag
SDP
Catalog No. NC0463185
Encompass_Preferred
Glovebag 17" x 17" x 11" with 11 1/2" equipment opening in the front between two integral turned in gloves.Made of 3mil low density polyethylene. Handy Lok clsoure.
Catalog No. NC0463185 Supplier Glas Col LLC Supplier No. 108D X1717H
Glovebag 17" x 17" x 11" with 11 1/2" equipment opening in the front between two integral turned in gloves.Made of 3mil low density polyethylene. Handy Lok clsoure.