Learn More
Medchemexpress LLC HY-17596 200mg Medchemexpress, Closantel CAS:57808-65-8 Purity:>98%
SDP
Catalog No. 502081996
Encompass_Preferred
Medchemexpress, HY-17596 200mg Closantel CAS:57808-65-8 Closantel is a halogenated salicylanilide with a potent anti-parasitic activity. Closantel is a potent and highly specific Onchocerca volvulus chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. Closantel inhibits the O. volvulus L3 to L4 molt of developing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1996 Supplier Medchemexpress LLC Supplier No. HY17596200MG
Medchemexpress, HY-17596 200mg Closantel CAS:57808-65-8 Closantel is a halogenated salicylanilide with a potent anti-parasitic activity. Closantel is a potent and highly specific Onchocerca volvulus chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. Closantel inhibits the O. volvulus L3 to L4 molt of developing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.