Learn More
Apexbio Technology LLC Closantel Sodium 61438-64-0 50mg
SDP
Catalog No. 502240797
Encompass_Preferred
Closantel Sodium (CAS 61438-64-0) is a small-molecule inhibitor targeting the KinA/Spo0F two-component signaling system It is designed to inhibit the KinA/Spo0F phosphotransfer interaction thereby interfering with bacterial signal transduction pathways Closantel Sodium exerts its biological activity primarily through inhibition of the KinA/Spo0F phosphotransfer In vitro studies demonstrate inhibitory activity with an IC50 value of approximately 3 8 M Based on these pharmacological properties Closantel Sodium holds research potential in antimicrobial studies particularly for investigating Gram-positive bacterial signaling mechanisms and antibiotic resistance pathways
This item is not returnable. View return policy
Catalog No. 50-224-0797 Supplier Apexbio Technology LLC Supplier No. B171550
Closantel Sodium (CAS 61438-64-0) is a small-molecule inhibitor targeting the KinA/Spo0F two-component signaling system It is designed to inhibit the KinA/Spo0F phosphotransfer interaction thereby interfering with bacterial signal transduction pathways Closantel Sodium exerts its biological activity primarily through inhibition of the KinA/Spo0F phosphotransfer In vitro studies demonstrate inhibitory activity with an IC50 value of approximately 3 8 M Based on these pharmacological properties Closantel Sodium holds research potential in antimicrobial studies particularly for investigating Gram-positive bacterial signaling mechanisms and antibiotic resistance pathways