Learn More
Medchemexpress LLC Closantel (sodium) | 61438-64-0 | 99.8% | 685.06 | 200 MG
SDP
Catalog No. 5000387137
Encompass_Preferred
Closantel sodium is a halogenated salicylanilide with potent anti-parasitic activity. It acts as a potent and highly specific *Onchocerca volvulus* chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. This compound inhibits the *O. volvulus* L3 to L4 molt of developing organisms. It is highly specific for filarial family chitinases compared to those from protozoans and the human chitinase, human chitotriosidase.
  • Potent anti-parasitic activity.
  • Highly specific inhibitor of filarial family chitinases.
  • Less effect on protozoans and human chitinase.
  • For research use only.
Catalog No. 50-003-87137 Supplier Medchemexpress LLC Supplier No. HY17596A200MG
Closantel sodium is a halogenated salicylanilide with potent anti-parasitic activity. It acts as a potent and highly specific *Onchocerca volvulus* chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. This compound inhibits the *O. volvulus* L3 to L4 molt of developing organisms. It is highly specific for filarial family chitinases compared to those from protozoans and the human chitinase, human chitotriosidase.
  • Potent anti-parasitic activity.
  • Highly specific inhibitor of filarial family chitinases.
  • Less effect on protozoans and human chitinase.
  • For research use only.