Learn More
BURKLE INC close-it control seal, 3.74x3.74 in. (95x95 mm), blue
Catalog No. 5000025601
Encompass_Preferred
Sampling from sacks, boxes, and bags with lances is common but creates holes that allow bulk goods to escape. The close-it seal prevents this easily.
close-it features a sandwich structure (aluminum/paper) that hermetically seals containers, acting as a vapor barrier to prevent hygroscopic powders from clumping.
Available in various colors with red, blue, green, yellow, black, or neutral (white) printing options.
Writable with ballpoint pen, pencil, or marker.
Its strong adhesive sticks firmly to most surfaces, including powder-coated, moist, or frozen ones where regular labels or tape fail.
Catalog No. 50-000-25601 Supplier BURKLE INC Supplier No. 53030015
Sampling from sacks, boxes, and bags with lances is common but creates holes that allow bulk goods to escape. The close-it seal prevents this easily.
close-it features a sandwich structure (aluminum/paper) that hermetically seals containers, acting as a vapor barrier to prevent hygroscopic powders from clumping.
Available in various colors with red, blue, green, yellow, black, or neutral (white) printing options.
Writable with ballpoint pen, pencil, or marker.
Its strong adhesive sticks firmly to most surfaces, including powder-coated, moist, or frozen ones where regular labels or tape fail.