Learn More
BURKLE INC close-it food & pharma control seal, 3.74x3.74 in. (95x95 mm), yellow
Supplier:  BURKLE INC 53032020
Encompass_Preferred
Close-it adhesive labels are specially designed for securely sealing sampled bags, meeting the highest hygiene standards required in the food and pharmaceutical industries. The food-grade adhesive complies with all US FDA regulations, ensuring safe and reliable sealing without contamination risks.
Featuring a durable aluminum foil barrier layer, Close-it labels effectively prevent bulk materials from leaking after sampling. Perfect for use with food products, pharmaceuticals, cosmetics, dietary supplements, and even frozen goods—any application demanding exceptional purity and safety.
Experience the trusted adhesive strength of the original Bürkle Close-it, now optimized for food and pharma applications in the US market, providing secure sealing to protect your samples and maintain product integrity.
Catalog No. 50-000-25558