Learn More
BURKLE INC close-it maxi control seal, 5.91x5.91 in. (150x150 mm), blue
Catalog No. NC2187663
Encompass_Preferred
Sampling from sacks, boxes, and bags with lances is common but creates holes that allow bulk goods to escape. The close-it seal prevents this easily.
close-it features a sandwich structure (aluminum/paper) that hermetically seals containers, acting as a vapor barrier to prevent hygroscopic powders from clumping.
Available in various colors with red, blue, green, yellow, black, or neutral (white) printing options.
Writable with ballpoint pen, pencil, or marker.
Its strong adhesive sticks firmly to most surfaces, including powder-coated, moist, or frozen ones where regular labels or tape fail.
This item is not returnable. View return policy
Catalog No. NC2187663 Supplier BURKLE INC Supplier No. 53031015
Sampling from sacks, boxes, and bags with lances is common but creates holes that allow bulk goods to escape. The close-it seal prevents this easily.
close-it features a sandwich structure (aluminum/paper) that hermetically seals containers, acting as a vapor barrier to prevent hygroscopic powders from clumping.
Available in various colors with red, blue, green, yellow, black, or neutral (white) printing options.
Writable with ballpoint pen, pencil, or marker.
Its strong adhesive sticks firmly to most surfaces, including powder-coated, moist, or frozen ones where regular labels or tape fail.