Learn More
BURKLE INC close-it tape food & pharma, WxL 95mm x 164.04 ft. (50 m), white
Supplier:  BURKLE INC 53032019
Encompass_Preferred
Besides the individually detachable close-it and close-it food & pharma closure labels, our repeatedly tested and proven control seal is now also available on a continuous roller. This helps you discover whether you require only a small piece, for example for closing a small hole, or for marking a specific point. Or whether you want to repair a long crack or a large hole quickly, reliably and easily. With its extremely strong adhesive strength, close-it tape holds on the most diverse bases, where the glue effect of normal adhesive tape quickly reaches its maximum capacity.
For universal use, we have kept the adhesive tape close-it tape in white with tear-off lines. It can be labeled with permanent ink and locks gas-proof. Available in the classic close-it design, as well as close-it tape food & pharma, with a food-compatible adhesive.
Catalog No. 50-000-25627