Learn More
Grainger Parker Close Pipe Nipple, 316 Stainless Steel, 3/4 in., 1 ea.
Catalog No. 502752576
Encompass_Preferred
These male close nipples are engineered for leak-free pipe connections in instrumentation and general industrial applications. Constructed from 316 stainless steel, these seamless fittings provide high-performance fluid and gas handling for various systems.
  • 316 stainless steel construction with bright annealed finish
  • Designed for 3/4 in. nominal pipe size
  • Threaded NPT male connections on both ends
  • Compatible with Schedule 80 pipe
  • Maximum operating pressure of 6,400 psi at 100°F
  • Operating temperature range of -425°F to +1,200°F
  • Meets ANSI B1.20.1, ANSI B31.1, and ANSI B31.3 standards
Catalog No. 50-275-2576 Supplier Grainger Supplier No. 14M043
These male close nipples are engineered for leak-free pipe connections in instrumentation and general industrial applications. Constructed from 316 stainless steel, these seamless fittings provide high-performance fluid and gas handling for various systems.
  • 316 stainless steel construction with bright annealed finish
  • Designed for 3/4 in. nominal pipe size
  • Threaded NPT male connections on both ends
  • Compatible with Schedule 80 pipe
  • Maximum operating pressure of 6,400 psi at 100°F
  • Operating temperature range of -425°F to +1,200°F
  • Meets ANSI B1.20.1, ANSI B31.1, and ANSI B31.3 standards