Learn More
Grainger Parker Pipe Nipple, Brass, 3/8 in Nominal Pipe Size, 1 in Length, 1,000 psi, 25/Pk.
Catalog No. 502771558
Encompass_Preferred
These precision-machined brass pipe nipples are designed for leak-free connections between pipes, fittings, and instruments. Constructed from durable C37700 brass, these nipples provide excellent resistance to corrosion from water, heat, acids, minerals, and peaty soils. They are suitable for industrial applications including air lines, water lines, and cooling lines.
  • Brass construction meets SAE J530 and SAE J531 standards
  • Designed for high-heat and industrial environments
  • Temperature range of -65°F to +250°F
  • Maximum operating pressure of 1,000 psi
  • Threaded on both ends with MNPT x MNPT connection type
  • Compatible with thread sealants such as PTFE tape
  • Designed for air and non-potable water applications
Catalog No. 50-277-1558 Supplier Grainger Supplier No. 13Y772
These precision-machined brass pipe nipples are designed for leak-free connections between pipes, fittings, and instruments. Constructed from durable C37700 brass, these nipples provide excellent resistance to corrosion from water, heat, acids, minerals, and peaty soils. They are suitable for industrial applications including air lines, water lines, and cooling lines.
  • Brass construction meets SAE J530 and SAE J531 standards
  • Designed for high-heat and industrial environments
  • Temperature range of -65°F to +250°F
  • Maximum operating pressure of 1,000 psi
  • Threaded on both ends with MNPT x MNPT connection type
  • Compatible with thread sealants such as PTFE tape
  • Designed for air and non-potable water applications