Learn More
Starna Cells Inc CLOSED SEAL CAP EA
SDP
Catalog No. NC9101382
Encompass_Preferred
Solid Screw Cap for screw top cells utilizing the GL14 caps
Catalog No. NC9101382 Supplier Starna Cells Inc Supplier No. GL14C
Solid Screw Cap for screw top cells utilizing the GL14 caps