Learn More
Sanisure Inc Closed System Graduated Cylinders forVolumetric Transfer
Catalog No. NC1849492
Encompass_Preferred
The SaniSure Closed System Graduated Cylinders provide the ability to make volumetric additionsfrom a closed system design directly to the process. The vented sealing plug on top of the cylinder allows full dispensing through the bottom port. These closed systems can be welded in-processthrough the attached weldable TPE tubing and be pumped out of the cylinder through a peristaltic pump utilizing the platinum cured silicone tubing segment. These pre-engineered solutions are available in 250mL, 500mL, 1L, and 2L volumes. SaniSure also offers full customization capabilities to meet your specific application needs
Catalog No. NC1849492 Supplier Sanisure Inc Supplier No. GC250SANG
The SaniSure Closed System Graduated Cylinders provide the ability to make volumetric additionsfrom a closed system design directly to the process. The vented sealing plug on top of the cylinder allows full dispensing through the bottom port. These closed systems can be welded in-processthrough the attached weldable TPE tubing and be pumped out of the cylinder through a peristaltic pump utilizing the platinum cured silicone tubing segment. These pre-engineered solutions are available in 250mL, 500mL, 1L, and 2L volumes. SaniSure also offers full customization capabilities to meet your specific application needs