Learn More
Grainger Closet Bend SS 3 in Socket x Push Fit
Catalog No. 502752231
Encompass_Preferred
The Blucher Closet Bend is a stainless steel closet bend for drainage applications. It has a 2 inch outlet size and is made from 304 stainless steel for corrosion resistance. The closet bend has a 90 degree angle and is designed to connect a toilet to the drainage system. It features a cleanout plug for access to clear clogs. The closet bend is lead-free and made in the USA. It meets ASME A112.19.2 standards.
Catalog No. 50-275-2231 Supplier Grainger Supplier No. 14N960
The Blucher Closet Bend is a stainless steel closet bend for drainage applications. It has a 2 inch outlet size and is made from 304 stainless steel for corrosion resistance. The closet bend has a 90 degree angle and is designed to connect a toilet to the drainage system. It features a cleanout plug for access to clear clogs. The closet bend is lead-free and made in the USA. It meets ASME A112.19.2 standards.