Learn More
Agilent Technologies Closing Mats, For 96-Well Plates, Silicone, Pre-Slit, 50/Pk.
Catalog No. NC2120146
Encompass
These silicone closing mats are designed for use with 96-well plates in chromatography applications, sample collection, and storage. The mats are pre-slit and feature round-well compatibility to ensure secure closure during laboratory procedures.
  • Designed for optimal use with high-performance liquid chromatography samplers
  • Pre-slit design for efficient sample access
  • Compatible with 96-well plate configurations
  • Supplied in a pack of 50
Catalog No. NC2120146 Supplier Agilent Technologies Supplier No. 50421389
These silicone closing mats are designed for use with 96-well plates in chromatography applications, sample collection, and storage. The mats are pre-slit and feature round-well compatibility to ensure secure closure during laboratory procedures.
  • Designed for optimal use with high-performance liquid chromatography samplers
  • Pre-slit design for efficient sample access
  • Compatible with 96-well plate configurations
  • Supplied in a pack of 50