Learn More
Med Vet International Skin Closure Strip Steri-Strip 1/2 X 4 inch Nonwoven Material Flexible Strip Tan, B1557, 6 Strips/Pack, 50 Packs/Box, 4 Boxes
Catalog No. 502831634
Encompass_Preferred
These Steri-Strip™ beige skin closures help to close wounds to minimize infections The strips cause less scarring than staples or sutures The material is porous to allow air circulation for better healing These sterile skin closure strips flex with the skin to ensure optimal protection They don't contain latex to minimize allergic reactions
Catalog No. 50-283-1634 Supplier Med Vet International Supplier No. 304148CS
These Steri-Strip™ beige skin closures help to close wounds to minimize infections The strips cause less scarring than staples or sutures The material is porous to allow air circulation for better healing These sterile skin closure strips flex with the skin to ensure optimal protection They don't contain latex to minimize allergic reactions