Learn More
ATCC Clostridioides difficile
Supplier:  ATCC 43598
Encompass
Clostridioides difficile strain 1470 was isolated in Belgium from the feces of an asymptomatic neonate. This whole-genome sequenced strain can be used in enteric disease research.
Catalog No. 50-238-3936