Learn More
ATCC Clostridioides difficile
Catalog No. 502383846
Encompass
This strain of Clostridioides difficile was isolated from a clinical specimen. It is toxigenic (toxinotype XXII) and was confirmed by PCR to have the cdtB and tcdA genes. This product has applications in enteric disease and infectious disease research.
Catalog No. 50-238-3846 Supplier ATCC Supplier No. BAA1814
This strain of Clostridioides difficile was isolated from a clinical specimen. It is toxigenic (toxinotype XXII) and was confirmed by PCR to have the cdtB and tcdA genes. This product has applications in enteric disease and infectious disease research.