Learn More
ATCC Clostridioides difficile
Supplier:  ATCC 17857
Encompass
Clostridioides difficile strain 870 is an anaerobic bacterium with applications in enteric disease research. This strain is toxinotype 0 with the presence of the tcdA and tctB genes confirmed by PCR.
Catalog No. 50-238-4315