Learn More
ATCC Clostridioides difficile
Supplier:  ATCC BAA1812
Encompass
This strain of Clostridioides difficile was isolated from a clinical specimen. It is toxigenic (toxinotype XII) and was confirmed by PCR to have the tcdA and tcdB genes. This product has applications in enteric disease and infectious disease research.
Catalog No. 50-238-2450