Learn More
ATCC Clostridioides difficile
Supplier:  ATCC BAA1803
Encompass
This strain of Clostridioides difficile was isolated from a clinical specimen. It is toxigenic (toxinotype IIIc) and was confirmed by PCR to have the cdtB, tcdA, and tcdB genes. This product has applications in enteric disease research.
Catalog No. 50-238-3672
 Restricted Item