Learn More
ATCC Clostridioides difficile
Supplier:  ATCC BAA1804
Encompass
This strain of Clostridioides difficile was isolated from a clinical specimen. It is toxigenic (toxinotype 0) and was confirmed by PCR to have the tcdA and tcdB genes. This product has applications in enteric disease research.
Catalog No. 50-238-3840
 Restricted Item