Learn More
ATCC Clostridioides difficile; Strain 5325
Catalog No. 502383532
Encompass
Clostridioides difficile strain 5325 is a whole-genome sequenced, anaerobic bacterium. This strain is toxigenic (toxinotype V) and is confirmed by PCR to have the cdtB, tcdA, and tcdB genes. This product has applications in enteric disease and infectious disease research.
Catalog No. 50-238-3532 Supplier ATCC Supplier No. BAA1875
Clostridioides difficile strain 5325 is a whole-genome sequenced, anaerobic bacterium. This strain is toxigenic (toxinotype V) and is confirmed by PCR to have the cdtB, tcdA, and tcdB genes. This product has applications in enteric disease and infectious disease research.