Learn More
ATCC Clostridioides difficile; Strain 3232
Catalog No. 502383852
Encompass
Clostridioides difficile strain 3232 was isolated from the feces of an adult with diarrhea. It is nontoxigenic and was confirmed by PCR to not have the tcdA and tcdB genes.
Catalog No. 50-238-3852 Supplier ATCC Supplier No. BAA1801
Clostridioides difficile strain 3232 was isolated from the feces of an adult with diarrhea. It is nontoxigenic and was confirmed by PCR to not have the tcdA and tcdB genes.