Learn More
ATCC Clostridioides difficile Strain LBM 0801058
Catalog No. 502380415
Encompass
Quality control strain for ChromID Clostridioides difficile agar.
Catalog No. 50-238-0415 Supplier ATCC Supplier No. BAA2155
Quality control strain for ChromID Clostridioides difficile agar.