Learn More
ATCC Clostridioides difficile Strain 630
Catalog No. 502382354
Encompass
Clostridioides difficile strain 630 is a whole-genome sequenced bacterium that was isolated in Switzerland from a clinical source. This bacterial anaerobe can be used in enteric disease research and is typed as toxinotype 0. Presence of the tcdA and tcdB genes was confirmed by PCR.
Catalog No. 50-238-2354 Supplier ATCC Supplier No. BAA1382
Clostridioides difficile strain 630 is a whole-genome sequenced bacterium that was isolated in Switzerland from a clinical source. This bacterial anaerobe can be used in enteric disease research and is typed as toxinotype 0. Presence of the tcdA and tcdB genes was confirmed by PCR.