Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Clostridium bifermentans derived from NCTC 506, KWIK-STIK
SDP

Catalog No. 23047150 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 KWIK-STIK
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-150 2 KWIK-STIK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-150 Supplier Microbiologics™ Supplier No. 01196P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Everything needed to grow reference cultures for QC testing is included in one device. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab. Available in a 2 pack, universally used for all types of microbiological control testing.

  • Biosafety Level: 2
  • 2 self-contained units of a single organism

Specifications

ATCC® NCTC 506
Biosafety Level 2
Certifications/Compliance ISO 11133
Content And Storage 2°C to 8°C
Format KWIK-STIK 2 Pack
Manufacturing Origin United States
Product Type Quality Control Organism
For Use With (Application) Media Testing, Microbial Identification
Packaging Type Foil Sleeves
Quantity 2 KWIK-STIK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.