Learn More
Zeptometrix Clostridium bifermentans Z176, Titered
SDP
Catalog No. 22158417 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-417 1 mL
1 options
Catalog No. 22-158-417 Supplier Zeptometrix Supplier No. 0804049
Add to Cart
Add to Cart
Live, titered microorganisms can be used to determine a limit of detection (LOD), in diagnostic assay development or cross reactivity studies. When used as a control for nucleic acid tests, the same protocols as those used to amplify clinical specimens should be employed.
Each frozen aliquot contains 1 mL of a pure, titered culture of Clostridium bifermentans. The identification of this organism was confirmed by 16S sequencing. The purity of the culture was monitored by Gram staining and by additional culturing. The titer was performed on a minimum of two aliquots after freezing. The freezing medium contains 15% glycerol as a cryoprotectant. Please see the Certificate of Analysis for the specific freezing medium used.
Specifications
Biosafety Level 1
Certifications/Compliance ISO 13485
Product Type Microorganism
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
Regulatory Status RUO
The purchase of infectious microorganisms from ZeptoMetrix requires a Material Transfer Agreement (MTA).