Learn More
ATCC Clostridium butyricum
Catalog No. 502382876
Encompass
Clostridium butyricum strain NCTC 7423 [IFO 13949, VPI 3266] was isolated from the intestine of a pig.
Catalog No. 50-238-2876 Supplier ATCC Supplier No. 19398
Add to Cart
Add to Cart
Clostridium butyricum strain NCTC 7423 [IFO 13949, VPI 3266] was isolated from the intestine of a pig.