Learn More
ATCC Albidiferax ferrireducens
Supplier:  ATCC BAA621
Encompass
Albidiferax ferrireducens strain T118 is a whole-genome sequenced bacterial type strain that was isolated from an aquifer in Oyster Bay in Virginia.
Catalog No. 50-238-1370