Learn More
Bioworld CLOSTRIDIUM DIFFICILE AGAR
Supplier:  Bioworld 306200191
Encompass_Preferred
Clostridium difficile Agar Used in the selective isolation and cultivation of Clostridium difficile; Prepared with the addition of blood, cycloserine, and cefoxitin ; pH: 7.4; Dehydrated Appearance: light beige, homogeneous powder; Prepa...
Catalog No. 50-254-294