Learn More
Alpco Ltd Clostridium difficile PCR Kit
SDP
Supplier:  Alpco Ltd 86CDFHU100
Encompass
Clostridium difficile PCR is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of Clostridium difficile (16s-rDNA) and Clostridium difficile toxin A (tcdA) / B (tcdB) genes in human stool samples and cultures. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
Catalog No. NC2284598