Learn More
ATCC Clostridium perfringens
Catalog No. 502382756
Encompass
Clostridium perfringens strain NCTC 8238 [281/50 is a bacterial anaerobe isolated from boiled salt beef. The cpa (Alpha toxin) and cpe (Enterotoxin) genes were confirmed to be present in this strain.
Catalog No. 50-238-2756 Supplier ATCC Supplier No. 12916
Add to Cart
Add to Cart
Clostridium perfringens strain NCTC 8238 [281/50 is a bacterial anaerobe isolated from boiled salt beef. The cpa (Alpha toxin) and cpe (Enterotoxin) genes were confirmed to be present in this strain.