Learn More
ATCC Clostridium perfringens
Supplier:  ATCC 19574
Encompass
Clostridium perfringens strain 43F4 is a bacterium that was isolated in Baltimore, Maryland, from human bile. This strain is toxigenic (Toxinotype A) and was confirmed by PCR to have the cpa, cpe, cpb2, and pfoA genes.
Catalog No. 50-238-3352