Learn More
ATCC Clostridium perfringens
Supplier:  ATCC 10543
Encompass
Clostridium perfringens strain BP6K is a whole-genome sequenced toxigenic bacterium that was confirmed to contain the cpa, cpb2, and pfoA genes. This anaerobe is known to produce large quantities of gas during growth, so cultivation in stoppered test tubes should be avoided or used with caution.
Catalog No. 50-238-4285