Learn More
ATCC Clostridium perfringens; genomic DNA
Catalog No. 502384636
Encompass
Genomic DNA isolated from Clostridium perfringens strain NCTC 8237 (ATCC 13124). This whole-genome sequenced product can be used in PCR and other molecular biology applications. The presence of the cpa and pfoA genes were confirmed by PCR.
Catalog No. 50-238-4636 Supplier ATCC Supplier No. 13124D5
Genomic DNA isolated from Clostridium perfringens strain NCTC 8237 (ATCC 13124). This whole-genome sequenced product can be used in PCR and other molecular biology applications. The presence of the cpa and pfoA genes were confirmed by PCR.