Learn More
ATCC Clostridium scindens
Catalog No. 502382631
Encompass
Clostridium scindens strain VPI 13733 is a whole-genome sequenced type strain that was isolated from feces. This bacterial culture produces 20 alpha-hydroxysteroid dehydrogenase, 7 beta-dehydrogenase, 7 alpha-dehydroxylase, bile acid, and the steroid desmolase.
Catalog No. 50-238-2631 Supplier ATCC Supplier No. 35704
Clostridium scindens strain VPI 13733 is a whole-genome sequenced type strain that was isolated from feces. This bacterial culture produces 20 alpha-hydroxysteroid dehydrogenase, 7 beta-dehydrogenase, 7 alpha-dehydroxylase, bile acid, and the steroid desmolase.