Learn More
ATCC Clostridium septicum; Strain Pasteur III
Supplier:  ATCC 12464
Encompass
Clostridium septicum strain Pasteur III (NCTC 547) is a whole-genome sequenced bacterial type strain. The culture can be used as a quality control strain for VITEK 2 Compact ANC card.
Catalog No. 50-238-3655