Learn More
ATCC Clostridium sporogenes
Supplier:  ATCC 19404
Encompass
Clostridium sporogenes strain SR 5 is a whole-genome sequenced bacterial strain that was isolated from a human with an acute case of gas gangrene. It is cited in USP <62> and <71> and has applications in media testing, quality control, sterility testing, and pharmaceutical and personal care.
Catalog No. 50-238-2437