Learn More
ATCC Clostridium sporogenes
Supplier:  ATCC 3584
Encompass
Clostridium sporogenes strain 388 is a whole-genome sequenced type strain that was isolated from soil. The bacterial culture can be used in sterility assurance, testing disinfectants, and quality control of ENDO-SPOR hydrogen peroxide sterilization.
Catalog No. 50-238-4192