Learn More
ATCC Clostridium sporogenes
Supplier:  ATCC 11437
Encompass
Clostridium sporogenes strain L.S. McClung 2006 is a heat-resistant bacterium that was isolated from a cotton plant. This whole-genome sequenced strain can be used in media testing, sterility testing, and quality control.
Catalog No. 50-238-4318