Learn More
ATCC Clostridium sporogenes; 6 x 0.3mL
Catalog No. 502381973
Encompass
ATCC 11437-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 11437™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-1973 Supplier ATCC Supplier No. 11437MINIPACK
ATCC 11437-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 11437™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.