Learn More
ATCC Clostridium sporogenes; genomic DNA
Catalog No. 502384325
Encompass
Genomic DNA isolated from Clostridium sporogenes strain L.S. McClung 2006 (ATCC 11437). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4325 Supplier ATCC Supplier No. 11437D5
Add to Cart
Add to Cart
Genomic DNA isolated from Clostridium sporogenes strain L.S. McClung 2006 (ATCC 11437). This whole-genome sequenced product can be used in PCR and other molecular biology applications.